Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HAI-2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA519360
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA519360 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA519360 Supplier Invitrogen™ Supplier No. PA519360
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with bovine and canine based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.

This gene encodes a transmembrane protein with two extracellular Kunitz domains that inhibits a variety of serine proteases. The protein inhibits HGF activator which prevents the formation of active hepatocyte growth factor. This gene is a putative tumor suppressor, and mutations in this gene result in congenital sodium diarrhea. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HAI-2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene SPINT2
Gene Accession No. O43291
Gene Alias AL024025; C76321; DIAR3; FLJ45571; HAI2; HAI-2; Hepatocyte growth factor activator inhibitor type 2; Kop; Kunitz-type protease inhibitor 2; PB; placental bikunin; serine peptidase inhibitor, Kunitz type 2; serine peptidase inhibitor, Kunitz type, 2; serine protease inhibitor, Kunitz type 2; serine protease inhibitor, Kunitz type, 2; Spint2; testicular tissue protein Li 183
Gene Symbols SPINT2
Host Species Goat
Immunogen Synthetic peptide sequence (PSAPRRQDSEDH) corresponding to the internal amino acids of SPINT2 (aa 111-123).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10653
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.