Learn More
Invitrogen™ HAI-2 Polyclonal Antibody

Description
HAI-2 Polyclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | HAI-2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 |
| Gene | SPINT2 |
| Gene Accession No. | O43291 |
| Gene Alias | AL024025; C76321; DIAR3; FLJ45571; HAI2; HAI-2; Hepatocyte growth factor activator inhibitor type 2; Kop; Kunitz-type protease inhibitor 2; PB; placental bikunin; serine peptidase inhibitor, Kunitz type 2; serine peptidase inhibitor, Kunitz type, 2; serine protease inhibitor, Kunitz type 2; serine protease inhibitor, Kunitz type, 2; Spint2; testicular tissue protein Li 183 |
| Gene Symbols | SPINT2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.