Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hairless Antibody (OTI1D9), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272023FR
Description
Hairless Monoclonal specifically detects Hairless in Human samples. It is validated for Western Blot.Specifications
Hairless | |
Monoclonal | |
DyLight 680 | |
PBS with 0.05% Sodium Azide | |
HR | |
Recombinant protein fragment corresponding to amino acids 946-1189 of human Hairless (NP_005135) produced in E.coli. | |
0.1 mL | |
55806.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI1D9 | |
Western Blot | |
ALUNC, AUHSA277165, FLJ98880, hairless (mouse) homolog, hairless homolog (mouse), MUHH, MUHH1, protein hairless | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction