Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hairy (Drosophila) Antibody (1/24) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$196.10 - $426.00
Specifications
Antigen | Hairy (Drosophila) |
---|---|
Clone | 1/24 |
Dilution | Western Blot : 1:100 - 1:2000, Immunohistochemistry, Chromatin Immunoprecipitation (ChIP) 1:10-1:500 |
Applications | Western Blot, Immunohistochemistry, ChIP Assay |
Classification | Monoclonal |
Description
Hairy (Drosophila) Monoclonal specifically detects Hairy (Drosophila) in Drosophila samples. It is validated for Western Blot, Chromatin Immunoprecipitation.Specifications
Hairy (Drosophila) | |
Western Blot : 1:100 - 1:2000, Immunohistochemistry, Chromatin Immunoprecipitation (ChIP) 1:10-1:500 | |
Monoclonal | |
Purified | |
Cellular Signaling, Developmental Biology, Epigenetics, Stem Cells | |
PBS with 0.02% sodium azide | |
IgG1 | |
Protein A purified | |
This antibody will stain Drosophila embryos. |
1/24 | |
Western Blot, Immunohistochemistry, ChIP Assay | |
Unconjugated | |
Mouse | |
Drosophila | |
Hairy gene product | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title