Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hairy (Drosophila) Antibody (1/24), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP276659F
Description
Hairy (Drosophila) Monoclonal antibody specifically detects Hairy (Drosophila) in Drosophila samples. It is validated for ImmunohistochemistrySpecifications
Hairy (Drosophila) | |
Monoclonal | |
FITC | |
PBS | |
Hairy gene product | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. | |
IgG1 |
Immunohistochemistry | |
2023-01-24 00:00:00 | |
Immunohistochemistry | |
Mouse | |
Protein A purified | |
RUO | |
Drosophila | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction