Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hairy (Drosophila) Antibody (3/30), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250417AF532
Description
Hairy (Drosophila) Monoclonal specifically detects Hairy (Drosophila) in Drosophila samples. It is validated for Western Blot.Specifications
Hairy (Drosophila) | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
Hairy gene product | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. |
Western Blot | |
3/30 | |
Western Blot | |
Mouse | |
Protein A purified | |
Cellular Signaling, Developmental Biology, Epigenetics, Stem Cells | |
Drosophila |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction