Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hairy (Drosophila) Antibody (3/30) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$196.10
Specifications
Antigen | Hairy (Drosophila) |
---|---|
Clone | 2024-03-30 00:00:00 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:100 - 1:2000 |
Applications | Western Blot |
Description
Hairy (Drosophila) Monoclonal antibody specifically detects Hairy (Drosophila) in Drosophila samples. It is validated for Western BlotSpecifications
Hairy (Drosophila) | |
1 mg/mL | |
Western Blot | |
Unconjugated | |
Mouse | |
Cellular Signaling, Developmental Biology, Epigenetics, Stem Cells | |
PBS | |
Hairy gene product | |
Protein A purified |
2024-03-30 00:00:00 | |
Western Blot 1:100 - 1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Drosophila | |
P14003, P14003 | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title