Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hairy (Drosophila) Antibody (3/30), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250417MFV450
Description
Hairy (Drosophila) Monoclonal antibody specifically detects Hairy (Drosophila) in Drosophila samples. It is validated for Western BlotSpecifications
| Hairy (Drosophila) | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Hairy gene product | |
| 0.1 mL | |
| Primary | |
| Store at 4C in the dark. |
| Western Blot | |
| 2024-03-30 00:00:00 | |
| Western Blot | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Drosophila | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction