Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hakai Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31
Specifications
Antigen | Hakai |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-156-3927
![]() |
Bethyl Laboratories, Inc
A302969AT |
20 μL |
Each of 1 for $151.31
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt.
Epithelial cell cadherin (CDH1; MIM 192090) is endocytosed as a consequence of tyrosine phosphorylation and ubiquitination. HAKAI is an E3 ubiquitin ligase (see UBE3A; MIM 601623) that mediates ubiquitination of the CDH1 complex.Specifications
Hakai | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
HAKAI, RNF188, E-cadherin binding protein E7, E3 ubiquitin-protein ligase Hakai, RING finger protein 188, c-Cbl-like protein 1, casitas B-lineage lymphoma-transforming sequence-like protein 1 | |
Between 441 and 491 | |
Primary | |
4° C | |
CBLL1 |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q75N03, Q9JIY2 | |
104836, 79872 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title