Learn More
Hakai Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302969A

Description
The recommended shelf life for this product is 1 year from date of receipt.
Epithelial cell cadherin (CDH1; MIM 192090) is endocytosed as a consequence of tyrosine phosphorylation and ubiquitination. HAKAI is an E3 ubiquitin ligase (see UBE3A; MIM 601623) that mediates ubiquitination of the CDH1 complex.Specifications
Hakai | |
Polyclonal | |
Unconjugated | |
CBLL1 | |
HAKAI, RNF188, E-cadherin binding protein E7, E3 ubiquitin-protein ligase Hakai, RING finger protein 188, c-Cbl-like protein 1, casitas B-lineage lymphoma-transforming sequence-like protein 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
104836, 79872 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q75N03, Q9JIY2 | |
CBLL1 | |
Between 441 and 491 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.