Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HAL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588922
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588922 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588922 Supplier Invitrogen™ Supplier No. PA588922
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MPRYTVHVRG EWLAVPCQDA QLTVGWLGRE AVRRYIKNKP DNGGFTSVDD AHFLVRRCKG LGLLDNEDRL EVALENNEFV EVVIEGDAMS PDFIPSQPEG VYLYSKYREP EKYIELDGDR LTTEDLVNLG KGRYKIKLTP TAEKRVQKSR EVIDSIIKEK TVVYGITTGF GKFARTVIPI NKLQELQVNL VRSHSSGVGK; Positive Samples: Mouse liver, Rat liver.

HAL is a cytosolic enzyme catalyzing the first reaction in histidine catabolism, the nonoxidative deamination of L-histidine to trans-urocanic acid. HAL defects cause histidinemia which is characterized by increased histidine and histamine and decreased urocanic acid in body fluids Histidine ammonia-lyase is a cytosolic enzyme catalyzing the first reaction in histidine catabolism, the nonoxidative deamination of L-histidine to trans-urocanic acid. Histidine ammonia-lyase defects cause histidinemia which is characterized by increased histidine and histamine and decreased urocanic acid in body fluids.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HAL
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 4.77 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene HAL
Gene Accession No. P21213, P35492, P42357
Gene Alias hal; HIS; histidase; histidine ammonia lyase; histidine ammonia-lyase; histidinemia; Hsd; HSTD; Huth; zgc:136980
Gene Symbols HAL
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15109, 29301, 3034
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.