Learn More
Impacto™ Half-Finger Anti-Impact Mesh Carpal Tunnel Gloves
Catalog No. 19400092
Click to view available options
Size:
X-Small
Small
Medium
Large
This item is not returnable. View return policy
This item is not returnable. View return policy
Gloves feature a unique channel that takes the pressure off of the median nerve, which alleviates symptoms of repetitive strain injuries such as carpal tunnel syndrome. VEP padding in the palm dissipates shock, while the half-finger design maximizes tactile feedback.
  • Half-finger enhances tactile feedback
  • Durable cowhide leather palm with VEP padding dissipates shock
  • Air mesh back increases ventilation
  • Gusseted fingers for comfort
Specifications
Back Material Nylon Mesh
Color Black
Material Cowhide Leather
Size X-Small
Finger Style Half
Glove Cuff Style Hook and Loop
Series ST8610
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.