Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Haliotis laevigata Perlwapin Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5117545
Description
This antibody has been tested in direct-ELISA.
Inhibits growth of calcium carbonate crystals. May inhibit growth of certain crystallographic planes in the mineral phase of nacre in the shell.
Specifications
Haliotis laevigata Perlwapin | |
Polyclonal | |
Unconjugated | |
Rabbit | |
Protein G | |
RUO | |
Mollusca | |
Antibody | |
IgG |
Western Blot | |
3 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Recombinant Haliotis laevigata Perlwapin protein (1-134aa). | |
100 μg | |
Primary | |
-20°C or -80°C if preferred | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction