Learn More
Eisco™ Halite Mineral Specimen
SCI_ED
Catalog No. S24691 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1 Ea.
10/Pk.
12/Pk.
Specimen Size:
2 to 3 cm
7 to 8 cm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Specimen Size
S24691 1 Ea. 2 to 3 cm
S26605 10/Pk. 2 to 3 cm
S24692 12/Pk. 2 to 3 cm
S26629 1 Ea. 7 to 8 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. S24691 Supplier Eisco™ Supplier No. ESNG0017
Add to Cart
Add to Cart
Halite Mineral Specimen ideal for teaching students about cleavage planes and how evaporate deposits form.
  • Soft, white to translucent mineral that grows into cleavable cubes
  • Example of cubic cleavage
  • When broken, it will continually break into smaller and smaller cubes
  • For use in classroom identification labs and exercises, correlates well to Next Generation Science Standards Evidence Statements
  • For simulating evaporite deposition
Specifications
Product Type Mineral
For Use With (Application) Study of cleavage planes and how evaporite deposits form in nature
Packaging Clear labeled and barcoded bag
Quantity 1 Ea.
Specimen Size 2 to 3 cm
Specimen Type Halite