Learn More
Eisco™ Halite Mineral Specimen
SCI_ED
Catalog No. S24692 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1 Ea.
10/Pk.
12/Pk.
Specimen Size:
2 to 3 cm
7 to 8 cm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Specimen Size
S24692 12/Pk. 2 to 3 cm
S24691 1 Ea. 2 to 3 cm
S26605 10/Pk. 2 to 3 cm
S26629 1 Ea. 7 to 8 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. S24692 Supplier Eisco™ Supplier No. ESNG0017PK12
Add to Cart
Add to Cart
Halite Mineral Specimen ideal for teaching students about cleavage planes and how evaporate deposits form.
  • Soft, white to translucent mineral that grows into cleavable cubes
  • Example of cubic cleavage
  • When broken, it will continually break into smaller and smaller cubes
  • For use in classroom identification labs and exercises, correlates well to Next Generation Science Standards Evidence Statements
  • For simulating evaporite deposition
Specifications
Product Type Mineral
For Use With (Application) Study of cleavage planes and how evaporite deposits form in nature
Packaging Clear labeled and barcoded bag
Quantity 12/Pk.
Specimen Size 2 to 3 cm
Specimen Type Halite