Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Halofuginone hydrobromide
GREENER_CHOICESDP

Catalog No. 199350 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-935-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-935-0 Supplier Tocris Bioscience™ Supplier No. 1993/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

High affinity competitive prolyl-tRNA synthetase inhibitor

Halofuginone hydrobromide is a high affinity competitive prolyl-tRNA synthetase inhibitor (Ki = 18.3 nM). Blocks expression of MMP2. Inhibits ECM invasion in vitro and lung metastasis by bladder cancer cells in mice. Inhibits the development of Th17-driven autoimmunity in a mouse model of multiple sclerosis by activating the amino acid response (AAR) pathway. Also antiparasitic.

Specifications

Chemical Name or Material (2R*,3S*)-7-Bromo-6-chloro-3-[3-(3-hydroxy-2-piperidinyl)-2-oxopropyl]-4-3H-quinazolinone hydrobromide
CAS 64924-67-0
Assay Percent Range >98%
Quantity 50 mg
Molecular Formula C16H17BrClN3O3.HBr
Formula Weight Observed MW: 495.59
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.