Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Haloperidol hydrochloride
GREENER_CHOICESDP

Catalog No. 0931100
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-311-00 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 09-311-00 Supplier Tocris Bioscience™ Supplier No. 0931/100

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Antagonist, partly D2 selective

Dopamine antagonist with selectivity for D2-like receptors (Ki values are 1.2, ∽ 7, 2.3, ∽ 80 and ∽ 100 nM for D2, D3, D4, D1 and D5 receptors respectively). Subtype-selective NMDA antagonist. Identified as targeting human host proteins that interact with SARS-CoV-2.

Chemical Identifiers

CAS 1511-16-6
Molecular Formula C21H24Cl2FNO2

Specifications

Quantity 100 mg
Formula Weight 412.33
Purity >99%
Molecular Weight (g/mol) 412.326
Chemical Name or Material Haloperidol hydrochloride
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.