Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Haloperidol, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18603095 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-603-095 1 kg Amber Glass Bottle
18-603-093 5 g Amber Glass Bottle
18-603-094 25 g Amber Glass Bottle
18-603-096 100 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-603-095 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. HA1031KGBL

HA103, 52-86-8, C21H23ClFNO2

Spectrum™ Chemical Haloperidol, USP is an antipsychotic medication with pharmacological effects similar to the phenothiazines. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 52-86-8
Molecular Formula C21H23ClFNO2
Molecular Weight (g/mol) 375.87
MDL Number MFCD00051423
InChI Key LNEPOXFFQSENCJ-UHFFFAOYSA-N
IUPAC Name 4-[4-(4-chlorophenyl)-4-hydroxypiperidin-1-yl]-1-(4-fluorophenyl)butan-1-one
SMILES OC1(CCN(CCCC(=O)C2=CC=C(F)C=C2)CC1)C1=CC=C(Cl)C=C1

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Loss on Drying 0.5%
Packaging Amber Glass Bottle
Quantity 1 kg
UN Number UN2811
Percent Purity 98 to 102%
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.