Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega HaloTag™ Biotin Ligand

Catalog No. prg8281
Click to view available options
Quantity:
30 μL

The HaloTag™ Technology enables covalent protein labeling and immobilization in vivo and in vitro.

  • Covalently Attach Fluorescent Labels or Affinity Tags to Membrane or Intracellular Proteins
  • Cell permeant TMR, Oregon Green(R), diAcFAM and Coumarin ligands readily cross the cell membrane for labeling intracellular proteins
  • Cell impermeant tags include Alexa Fluor(R) 488 and 660 for quick cell surface labeling

Specifications

Concentration 5 mM
Content And Storage -30°C to -10°C
Quantity 30 μL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.