Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega HaloTag™ Mammalian Protein Purification System

Catalog No. PRG6790 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Product Type:
Purification System
Purification and Detection System
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
PRG6790 Purification System
PRG6795 Purification and Detection System
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRG6790 Supplier Promega Supplier No. G6790
Add to Cart
Add to Cart

Includes

HaloLink Resin, HaloTag TMRDirect Ligand, Protease Inhibitor Cocktail (50X), HaloTEV Protease, Spin Columns

HaloTag™ Mammalian Protein Purification System is an optimized kit for purification of HaloTag™ fusion proteins from mammalian cell culture lysates. HaloTag™ Mammalian Protein Detection and Purification System also includes HaloTag™ TMRDirect Ligand.

  • Recover Highly Pure Recombinant Protein from Mammalian Cell Cultures, Even at Low Expression Levels
  • HaloLink™ Resin covalently binds >7mg/mL of HaloTag(R) fusion protein (10X more capacity than FLAG(R))
  • Easy to scale up or down to obtain milligram + quantities of protein
  • Choose from purification, purification + detection or sample purification + detection kits

Specifications

Product Type Purification System
Quantity 1 Ea.
Includes HaloLink™ Resin, Protease inhibito r cocktail, 50X, HaloTEV Protease, spin column
Content And Storage COMPONENTS AT DIFFERENT TEMPS
Column Type Affinity Column
For Use With (Application) Purification of HaloTag™ fusion proteins from mammalian cell culture lysates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.