Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Halotyrosine Monoclonal Antibody (BTK-94C)
GREENER_CHOICE

Catalog No. FERMA549046
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERMA549046 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERMA549046 Supplier Invitrogen™ Supplier No. MA549046
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Halotyrosine, clone BTK-94C is recommended for detection of bromotyrosine modified proteins.

Halotyrosine is a type of protein modification that results from the addition of a halogen element, usually bromine or chlorine, to a tyrosine residue. This process is often a result of immune cell actions or oxidative stress. Halotyrosine residues are commonly found in halogenated organic compounds, which are natural products found in many living organisms. Research has shown that total bromotyrosine levels, a type of halotyrosine residue, are elevated in individuals with asthma but decrease in response to anti-inflammatory medications.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Halotyrosine
Applications Inhibition Assays, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone BTK-94C
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA and 0.02% sodium azide
Gene Alias Halo tyrosine
Host Species Mouse
Immunogen Bromotyrosine structural mimic conjugated to KLH.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Tag
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgM
Target Molecule Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.