Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Halt™ Phosphatase Inhibitor Cocktail, 1 mL PROMO
Click to view available options
:
Thermo Scientific™ Halt™ Phosphatase Inhibitor Cocktail, 1 mL
Description
- Halt Phosphatase Inhibitor Cocktail protects phosphoproteins from serine/threonine phosphatases, as well as protein tyrosine phosphatases (PTPs) during cell lysis
- The cocktail contains a mixture of four inhibitors of broad specificity including sodium fluoride, sodium orthovanadate, sodium pyrophosphate, and beta-glycerophosphate
- Conveniently packaged in a 100X format, the cocktail is stable at 4°C and is suitable for use with any cell or tissue extract
- The cocktail preserves protein phosphorylation in immunoprecipitation and kinase assays
- Phosphorylation and dephosphorylation comprise a molecular on/off switch that regulates many key biological pathways within the cell including signal transduction, cell division, and apoptosis
- To effectively study phosphorylation activation states, one must inactivate phosphatases to prevent their uncontrolled activity upon cell lysis
Additional Features
- Devoid of protease inhibitors;– To protect against both proteases and phosphatases, use Halt Protease and Phosphatase Inhibitor Cocktail (78420)
- Dialyzable;–;Either dialyze or desalt sample to effectively remove inhibitors from cell extracts before performing procedures that are discovered to be incompatible with cocktail components
- Supplied as a 100X concentrate;–;effective when used at a final concentration of 1X; however, samples that contain particularly high levels of phosphatase activity may require a final concentration of 2-3X
- Compatibility –;compatible with most detergent-based cell lysis reagents, including all Thermo Scientific Pierce Cell Lysis Solutions
- All-in-one format;–;inhibits both serine-threonine phosphatases and protein tyrosine phosphatases
- Compatible with popular protein assays;–;works with standard protein assays, including BCA and coomassie (Bradford) assays
- Convenient storage;–;refrigerator storage (2 to 8°C) saves valuable space in the freezer and allows for immediate use
Specifications
Specifications
| Product Type | Phosphatase Inhibitor Cocktail |
| Content And Storage | Upon receipt store product at 4°C. Do not freeze. |
| Form | Liquid |
| Product Line | Halt™ |
| Reagent Type | Phosphatase Inhibitor |
| Inhibitors | Phosphatase Inhibitor |
| Quantity | 1 mL |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction