Learn More
Thermo Scientific™ Halt™ Protease Inhibitor Cocktail, EDTA-Free (100X), 1 mL PROMO
Description
Halt Protease Inhibitor Cocktails are 100X solutions containing optimized concentrations of six broad-spectrum protease inhibitors stabilized in high-quality dimethylsulfoxide (DMSO). The cocktails effectively inhibit serine-proteases, cysteine-proteases, aspartic acid-proteases and aminopeptidases that are typically present in cellular lysate samples. The identity and concentration of each component is fully disclosed, leaving no uncertainties about variables and compatibility with experiments. Regular and EDTA-free formulations provide for inhibition of metalloproteases and compatibility with 2D electrophoresis, respectively. The stable, concentrated liquid cocktails are easy to use, and the several different package sizes make Halt Protease Inhibitor Cocktails exceptionally convenient for either small-scale or large-scale uses.
Highlights:
Better than tablets – the refrigerator-stable, 100X liquid format is more effective and easier to use than individual inhibitors or other formats; just pipette exactly the amount you need
No proprietary ingredients – fully disclosed formulation contains AEBSF, aprotinin, bestatin, E-64, leupeptin and pepstatin A in DMSO
Multiple package sizes – choose among several package sizes, including single-use microtubes that keep protease inhibitors fresh and reduce the risk of reagent contamination
Two popular formulations – regular formulation includes a separate vial of EDTA solution for optional metalloprotease inhibition; EDTA-free formulation ensures compatibility with isoelectric focusing
Lysis buffer compatible – use with Thermo Scientific Pierce™ Cell Lysis Buffers or nearly any other commercial or homemade detergent-based lysis reagent
Includes:
Protease inhibitor cocktails with EDTA include separate 0.5M EDTA solution.
Specifications
Specifications
| Product Type | Protease Inhibitor Cocktail |
| Content And Storage | Upon receipt store at 4°C |
| Form | Liquid |
| Product Line | Halt™ |
| Buffer | Lysis Buffer |
| Reagent Type | Protease Inhibitor |
| Inhibitors | Protease Inhibitor |
| Quantity | 1 mL |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.