Learn More
Thermo Scientific™ Halt™ Protease and Phosphatase Inhibitor Cocktail (100X), 1 mL PROMO
Description
This broad-spectrum Halt Protease and Phosphatase Inhibitor Cocktail is specifically optimized to protect proteins from degradation during extraction and purification. The cocktail contains inhibitors against the major classes of proteases and phosphatases, targeting aminopeptidases, cysteine and serine proteases and serine/threonine and protein tyrosine phosphatases. The 100X cocktails, available in regular and EDTA-free formulations and several package formats, are refrigerator-stable liquids that are very easy to use for nearly any sample or experimental scale.
Highlights:
Complete protection all-in-one cocktail contains both protease and phosphatase inhibitors
Multiple package sizes choose from four convenient packages sizes to match nearly any scale
Convenient the 100X liquid formulation is much easier and more effective to use than individual inhibitors and other formats
Easy storage aqueous-based format allows for convenient cold-room storage without freezing, by contrast with DMSO-based cocktails
Compatible can be used in mass spectrometry (MS) because it does not contain AEBSF, which can cause peaks to shift
Validated fully compatible with Thermo Scientific Pierce™ Cell Lysis Buffers
Includes:
Formulations with and without supplementary vial of EDTA for metalloprotease inhibition, both of which are offered in four package sizes.
Specifications
Specifications
Product Type | Protease and Phosphatase Inhibitor Cocktail |
Content And Storage | Upon receipt store at 4°C |
Form | Liquid |
Product Line | Halt™ |
Buffer | Lysis Buffer |
Reagent Type | Protease Inhibitor, Phosphatase Inhibitor |
Inhibitors | Protease Inhibitor, Phosphatase Inhibitor |
Quantity | 1 mL |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.