Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Halyard Health Purple Nitrile™ Gloves

Catalog No. p-5159535
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
This item is not returnable. View return policy

Catalog No. 23-500-117

Supplier: Halyard Health 55080H

Only null left
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Offer the best barrier protection against bacteria, viruses, and a broad range of chemicals

  • Textured fingertips enhance grip while providing excellent tactile sensitivity
  • Latex-free and powder-free with no irritating perfume odor
  • Ambidextrous
  • Available as both sterile and non-sterile

Certifications

510K approved for medical use

Specifications

Color Purple
Disposable Single-use
Hand Ambidextrous
Latex Free Yes
Material Nitrile
Powdered Powder Free
Size X-Small
Thickness (English) 5.9 mil
Finger Style Textured
Glove Cuff Style Beaded/Rolled
Model 55080H
Tensile Strength 21
Length (English) 9.5 in.
Length (Metric) 24.13 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.