Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Halyard Health Universal Precautions Lab Coats

Catalog No. 23500173
Click to view available options
Color:
Blue
White
Size:
Small
Medium
Large
X-Large
2X-Large
Length (Metric):
105.4 cm
116.8 cm
118.1 cm
119.4 cm
This item is not returnable. View return policy

Catalog No. 23-500-173

Supplier: Halyard Health 10044

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Protective tri-layer fabric offers resistance to blood strikethrough at various levels of exposure

  • For low to moderate exposure to fluids or chemicals
  • Traditional collar and knit cuffs
  • Six snaps
  • Three reinforced pockets
  • Back vent
  • Neck loop for hanging
  • Size/name label
  • Available in colors blue and white

Specifications

Size 2X-Large
Material SMS Fabric
Color White
Cuff Style Knit
Description 3-layer SMS fabric
Size (Metric) Chest 81.45 cm
Length (Metric) 119.4 cm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.