Learn More
Hamilton™ Chemically Inert Manual Valves
Valves have a fluid path containing PTFE and CTFE
$140.00 - $166.00
Specifications
Material CTFE, PTFE
View More Specs
Products 3
Catalog Number Mfr. No. Type Price Quantity & Availability  
Catalog Number Mfr. No. Type Price Quantity & Availability  
14-815-354
View Documents
Hamilton™
86727
3-3 Manual Valve
Each for $166.00
Only null left
Add to Cart
14-819-280
View Documents
Hamilton™
86725
1-1 Manual Valve
Each for $140.00
Only null left
Add to Cart
14-819-281
View Documents
Hamilton™
86728
3-2 Manual Valve
Each for $166.00
Only null left
Add to Cart
Description
Description
The ports on the valve are 1/4-28 threads and come in different flow paths.
Specifications
Specifications
CTFE, PTFE
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.