Learn More
Hamilton™ Gastight™ Syringe, 500 μL, Sold by MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsSpecifications
Specifications
| Material | Borosilicate Glass (Barrel), PTFE (Plunger), PTFE (Tip) |
| Product Type | Gastight™ Syringe |
| Max. Pressure | 700 psig (48.3 bar) |
| Model | 1750 TLL |
| Quantity | 1 Ea. |
| Volume (Metric) | 500 μL |
| Product Line | Hamilton |
| Needle Point Style | No Needle Included |
| Connection Type | Luer Lock with Slots |
GASTIGHT is a registered trademark of Hamilton Co.