Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hamilton™ PRP™-3 Reversed Phase HPLC Columns

Catalog No. p-51763 Shop All Hamilton Company Products
Change view
Click to view available options
Length (Metric):
150 mm
250 mm
305 mm
Diameter (Metric):
4.1 mm
7 mm
10 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric)
14-816-101 250 mm 10 mm
14-816-39 150 mm 4.1 mm
14-816-67 305 mm 7 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 14-816-101 Supplier Hamilton™ Supplier No. 79526
Add to Cart
Add to Cart
This item is not returnable. View return policy

PRP-3 polymeric 300 Å columns for gradient separations of peptides and proteins

PRP-3 polymeric reversed phase columns, packed with a poly(styrene-divinyl)benzene support, last longer than silica-based columns. Support is pressure stable up to 5,000 psi and crosslinked to prevent shrinkage or swelling when the mobile phase is changed. The support is entirely polymeric and there are no C18 groups to hydrolyze, column lifetime is longer.

Superior pH Stability

  • The inherent stability of PRP-3 columns allows their use for any protein purification method from pH 1 to 13. PRP-3 columns allow you to analyze protein purity at low and high pH extremes.
  • Protein Recovery
    • The 300Å pores on the PRP-3 support help ensure good recovery (>90%) of your protein. The highly inert poly(styrene-divinylbenzene) packing enhances protein recovery because there are no silanol groups on the support to cause irreversible protein adsorption
  • Wide Applicability
    • Easy to use and ideal for almost any protein separation, including pH conditions from 1 to 13, with organic modifier (0-100%) of virtually any type from routine to the unusual
  • Scaleup
    • A selection of column sizes are available from analytical to semi-prep and preparative scale. Sample scaleup is easy because the PRP-3 packing is consistent from analytical to preparative columns.
    • The short analytical (50mm) column is well suited for the gradient elution of high molecular weight proteins. The longer (150mm) column is well suited for smaller proteins.

    Product Features:

    • Particle size: 10μm and 12-20μL
    • Eight column internal diameters: 2.1 to 21.2μL
    • Two column materials: 316 stainless steel and PEEK
    • Analytical and semiprep/preparative guard columns
    • Custom hardware dimensions available

    Specifications

    Column Format Preparative Column
    Diameter (Metric) 10 mm
    Length (Metric) 250 mm
    Max. Pressure 5000 psi (345 bar)
    Particle Size 10 μm
    pH 1 to 13
    Pore Size 300 Å
    Temperature 5°C to 85°C
    USP Type L21
    Stationary Phase PRP-3
    Packing Material PSDVB (Polystyrene Divinylbenzene)
    Column Type Reversed Phase
    Phase Reversed Phase
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.