Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hamilton™ PRP-X100 Anion Exchange HPLC Columns: PEEK

Catalog No. 13622550 Shop All Hamilton Company Products
Change view
Click to view available options
Particle Size:
5 μm
10 μm
Flow Rate:
1 mL/min
8 mL/min
Column Format:
150 x 2.1 mm L x ID
150 x 4.6 mm
150 x 4.6 mm L x ID
250 x 4.6 mm L x ID
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Particle Size Flow Rate Column Format
13-622-550 5 μm 1 mL/min 150 x 2.1 mm L x ID
14-816-71 10 μm 8 mL/min 150 x 4.6 mm
13-622-555 5 μm 1 mL/min 150 x 4.6 mm L x ID
13-622-556 5 μm 1 mL/min 250 x 4.6 mm L x ID
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 13-622-550 Supplier Hamilton™ Supplier No. 79852
Only null left
Add to Cart
Add to Cart

Polymeric column packing for separation of inorganic and organic anions from 10 to 500 ppm

Easy Ion Determination

  • PRP-X100 ion chromatography columns are the easy way to separate inorganic ions.These columns can be used with any existing HPLC or ion chromatograph to easily determine anions in almost any sample matrix including air, water and soil.
Better than Wet Chemical Methods
  • Ion chromatography greatly reduces sample pretreatment and improves the accuracy and precision of results when using wet chemical or colorimetric methods
High Sensitivity
  • Use to separate ions at concentrations from 1 ppm to 1,000 ppm without suppression or elaborate preconcentration techniques
Long Column Life
  • Highly inert polymeric support resists chemical attack from organic solvents and aqueous buffers (0-100% aqueous or organic; pH 1 to 13), effectively lengthening column life. If column performance should deteriorate (peak broadening and a loss of symmetry), the regeneration protocol will usually return chromatography to its original state.
Analyze Both Difficult and Common Anions
  • Easily separate difficult anions such as cyanide, borate and silicate at high pH (11.5). The polymeric packing is stable from pH 1 to 13, so a single column can be used for the analysis of both common and difficult anions.
Easy Detection and Common Eluents
  • Conductivity detection works by measuring the increased conductance of the eluting sample ions relative to the background conductance

Specifications

Column Format 150 x 2.1 mm L x ID
Diameter (Metric) 2.1 mm
Flow Rate 1 mL/min
For Use With Analysis of Fluoride, Carbonate, Chloride, Nitrite, Bromide, Nitrate, Phosphate, Sulfate and Other Ions
Length (Metric) 150 mm
Max. Pressure 5000 psi (345 bar)
Mobile Phase Compatibility pH 1 to 13, 0 to 100% aqueous, organic modifier
Particle Size 5 μm
pH 1 to 13
Pore Size 100 Å
Temperature 5°C to 60°C
USP Type L47
Stationary Phase PRP-X100
Packing Material PSDVB/Trimethylammonium
Column Type Ion Exchange
Phase Ion Exchange
Material PEEK
Type Column
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.