Learn More
Restek™ Hamilton™ Micro-Volume Positive Displacement Syringe, 2.0μL
SDP
Catalog No. 06712341 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Volume (Metric):
2.0 μL
Catalog No. Volume (Metric)
06-712-341 2.0 μL
1 options
Catalog No. 06-712-341 Supplier Restek™ Supplier No. 24551
Use these syringes for accurate, high precision injections. Restek™ Hamilton™ Micro-Volume Positive Displacement Syringes are designed to contain the sample in the needle with full plunger displacement, ensuring no dead volume.
  • High precision and accuracy down to 0.1uL.
  • Seal between the plunger and the needle can be tightened for high-pressure injection.
  • Replaceable needle/plunger kits and barrels (available on request).
Specifications
Needle Gauge 25 G
Length (English) 2.75 in.
Length (Metric) 70 mm
Volume (Metric) 2.0 μL
Needle Point Style 2
Connection Type Knurled Hub