Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poltex Hamilton Tube Rack Stabilizer
SDP

Catalog No. p-7163852
Click to view available options
Size:
1 slot
2 slot
3 slot

Stabilizes extractor tube strip/racks when loading/unloading.

Durable and cleanable PETG construction

Specifications

Autoclavable Not Autoclavable
Color Clear
Holds Blood Tube Sample Strip
Material PETG
Positions 1
Sterility Non-sterile
For Use With (Equipment) Qiasymphony extractor racks, hamilton racks
Quantity 1
Size 1 slot
Length (Metric) 482 mm
Width (Metric) 108 mm
Height (Metric) 25 mm
Weight (Metric) 150 g
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.