Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bcl2l11, Hamster, Clone: Ham151-149, Abnova™

Catalog No. 89113180 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-180 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-180 Supplier Abnova Corporation Supplier No. MAB7553

Hamster monoclonal antibody raised against native Bcl2l11.

Specifications

Antigen Bcl2l11
Applications Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone Ham151-149
Conjugate Unconjugated
Description BCL2-like 11 (apoptosis facilitator)
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene Bcl2l11
Gene Alias 1500006F24Rik, Bim, Bod
Gene Symbols Bcl2l11
Host Species Hamster
Immunogen Native purified Bcl2l11 from mouse BimL (w/o transmembrane region) and Bcl-xL (w/o transmembrane region)-6xHis tag, purified from Hi5 cells.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12125
Test Specificity This antibody reacts with mouse Bim, a 19-24kDa pro-apoptotic protein (Bcl-2 family) which regulates immunological responses.
Target Species Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.