Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bcl3, Hamster, Clone: 150-3.5, Abnova™

Catalog No. 89110879 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-879 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-879 Supplier Abnova Corporation Supplier No. MAB2192
Add to Cart
Add to Cart

Hamster monoclonal antibody raised against recombinant Bcl3.

Specifications

Antigen Bcl3
Applications ELISA, Western Blot
Classification Monoclonal
Clone 150-3.5
Conjugate Unconjugated
Description Hamster monoclonal antibody raised against recombinant Bcl3.
Dilution Western Blot (2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4
Gene Bcl3
Gene Alias AI528691/Bcl-3
Gene Symbols Bcl3
Host Species Hamster
Immunogen Recombinant His fusion protein corresponding to mouse Bcl3.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12051
Test Specificity This antibody recognizes human and mouse Bcl-3.
Target Species Human, Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.