Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hamster anti-Bcl3, Clone: B325H, Abnova™

Catalog No. 89111649 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-649 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-649 Supplier Abnova Corporation Supplier No. MAB3894

Specifications

Antigen Bcl3
Applications ELISA, Western Blot
Classification Monoclonal
Clone B325H
Conjugate Unconjugated
Formulation In PBS, pH 7.4
Gene Symbols Bcl3
Host Species Hamster
Immunogen Recombinant 6His fusion protein corresponding to mouse Bcl3.
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 12051
Target Species Human, Mouse
Content And Storage Store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.