Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD152 Hamster anti-Mouse, APC-R700, Clone: UC10-4F10-11, BD

Catalog No. BDB565778 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB565778 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB565778 Supplier BD Biosciences Supplier No. 565778
Add to Cart
Add to Cart

Hamster Monoclonal Antibody

The UC10-4F10-11 monoclonal antibody specifically binds to CD152, which is also known as Cytotoxic T-lymphocyte-associated antigen 4 (CTLA-4). CD152 is expressed on activated T lymphocytes 2-3 days after stimulation through T cell receptor. CTLA-4 has significant similarity to CD28 in amino acid sequence, structure, and genomic organization. Furthermore, CD152 and CD28 share common B7 family counter-receptors. Unlike CD28, CD152 expression appears to be restricted to activated T cells and CD25+CD4+ regulatory T (Treg) cells. Whereas CD28 delivers a costimulatory signal required for T-cell activation, CTLA-4 is a negative regulator of cell-mediated immune responses. CD152 may play roles in induction and/or maintenance of immunological tolerance, regulation of protective immunity, and autoimmune responses, and regulation of some aspects of thymocyte maturation. This hamster mAb to a mouse leukocyte antigen does not cross-react with rat leucocytes.

This antibody is conjugated to BD Horizon APC-R700, which has been developed exclusively by BD Biosciences as a better alternative to Alexa Fluor™ 700. APC-R700 excites and emits at similar wavelengths to Alexa Fluor™ 700 yet exhibits significantly improved brightness. This dye can be excited by the red laser and detected with the same filter set as Alexa Fluor™ (eg, 730/45-nm filter).

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD152
Applications Flow Cytometry
Classification Monoclonal
Clone UC10-4F10-11
Concentration 0.2mg/mL
Conjugate APC-R700
Description Ctla4; CTLA-4; Cytotoxic T-lymphocyte-associated protein 4; Ly-56
Formulation Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide.
Host Species Hamster
Immunogen Mouse CTLA-4 IgG2a Fusion
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.