Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cd247, Hamster, FITC, Clone: H146-968, Abnova™

Catalog No. 89113805 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-805 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-805 Supplier Abnova Corporation Supplier No. MAB9821

Hamster monoclonal antibody raised against synthetic peptide of Cd247.

Specifications

Antigen Cd247
Applications Flow Cytometry
Classification Monoclonal
Clone H146-968
Conjugate FITC
Description CD247 antigen
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene Cd247
Gene Alias 4930549J05Rik, AW552088, CD3-eta, CD3-zeta, CD3zeta, Cd3, Cd3h, Cd3z, T3z, TCRk, Tcrz
Gene Symbols Cd247
Host Species Hamster
Immunogen A synthetic peptide corresponding to amino acids 151-164 of mouse Cd247.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12503
Test Specificity The hamster antibody H146-968 reacts with CD247 (CD3 zeta chain), which is a component of TCR/CD3 complex expressed on T cells.
Target Species Human, Mouse
Content And Storage Store in the dark at 4°C. Avoid prolonged exposure to light. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.