Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD27/Cd27, Hamster, Clone: LG.3A10, Abnova™

Catalog No. 89109275 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-275 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-275 Supplier Abnova Corporation Supplier No. MAB11067

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Armenian hamster monoclonal antibody raised against CD27/Cd27.

Specifications

Antigen CD27/Cd27
Applications Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone LG.3A10
Conjugate Unconjugated
Description Armenian hamster monoclonal antibody raised against CD27/Cd27.
Dilution The optimal working dilution should be determined by the end user.
Formulation In aqueous buffer (0.09% sodium azide)
Gene Alias MGC20393/S152/T14/TNFRSF7/Tp55
Gene Symbols CD27
Host Species Hamster
Immunogen Human CD27 and mouse Cd27.
Purification Method Affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 939/21940
Target Species Human, Mouse
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.