Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cd28, Hamster, Clone: 37.51, Abnova™

Catalog No. 89112734 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-734 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-734 Supplier Abnova Corporation Supplier No. MAB6206
Add to Cart
Add to Cart

Hamster monoclonal antibody raised against Cd28.

Specifications

Antigen Cd28
Applications Flow Cytometry, Immunoprecipitation
Classification Monoclonal
Clone 37.51
Conjugate Unconjugated
Description Hamster monoclonal antibody raised against Cd28.
Dilution The optimal working dilution should be determined by the end user.
Formulation In 100mM BBS, pH 8.2
Gene Cd28
Gene Symbols Cd28
Host Species Hamster
Immunogen C57BL/6 mouse T-cell lymphoma EL-4.
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12487
Test Specificity CD28 costimulatory receptor, Mr 40 KDa.
Target Species Mouse
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.