Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cd69, Hamster, FITC, Clone: H1.2F3, Abnova™

Catalog No. 89112414 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-414 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-414 Supplier Abnova Corporation Supplier No. MAB5742

Hamster monoclonal antibody raised against native Cd69.

Specifications

Antigen Cd69
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation
Classification Monoclonal
Clone H1.2F3
Conjugate FITC
Description Hamster monoclonal antibody raised against native Cd69.
Dilution Flow Cytometry (1 ug/106 cells) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene Cd69
Gene Alias 5830438K24Rik/AI452015/AIM/VEA
Gene Symbols Cd69
Host Species Hamster
Immunogen Native purified Cd69 from Y245 mouse dendritic epidermal T cell line.
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12515
Test Specificity Mouse CD69/Very Early Activation Antigen (VEA), Mr 85 KDa (unreduced).
Target Species Mouse
Content And Storage Store in the dark at 4°C. Do not freeze.
Avoid prolonged exposure to light.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG λ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.