Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cd79a/Cd79b, Hamster, FITC, Clone: HM79, Abnova™

Catalog No. 89113806 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-806 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-806 Supplier Abnova Corporation Supplier No. MAB9822

Hamster monoclonal antibody raised against native Cd79a/Cd79b.

Specifications

Antigen Cd79a/Cd79b
Applications Flow Cytometry
Classification Monoclonal
Clone HM79
Conjugate FITC
Formulation In PBS, pH 7.4 (0.02% BSA, 0.09% sodium azide)
Gene Symbols Cd79a
Host Species Hamster
Immunogen Native purified Cd79a/Cd79b dimers from WEHI231 cells.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12518;15985
Test Specificity The hamster monoclonal antibody HM79 recognizes an extracellular epitope of mouse Cd79b (CD79 beta, Ig beta), a component of B cell receptor (BCR) complex.
Target Species Mouse
Content And Storage Store in the dark at 4°C. Avoid prolonged exposure to light. Do not freeze.
Isotype IgG2
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.