Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cd80, Hamster, Clone: 16-10A1, Abnova™

Catalog No. 89108424 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-108-424 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-108-424 Supplier Abnova Corporation Supplier No. MAB10177
Add to Cart
Add to Cart

Hamster monoclonal antibody raised against Cd80.

Specifications

Antigen Cd80
Applications Flow Cytometry, Functional Assay, Immunohistochemistry (Frozen), Immunoprecipitation
Classification Monoclonal
Clone 16-10A1
Conjugate Unconjugated
Description Hamster monoclonal antibody raised against Cd80.
Dilution Flow Cytometry (6 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene Cd80
Gene Alias B7-1/B7.1/Cd28l/Ly-53/Ly53/MIC17/TS/A-1
Gene Symbols Cd80
Host Species Hamster
Immunogen Mouse Cd80 transfected CHO cell line.
Purification Method Protein A purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12519
Target Species Canine, Mouse
Content And Storage Store at 4°C. Do not freeze.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.