Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cd81, Hamster, PE, Clone: 2F7, Abnova™

Catalog No. 89112463 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-463 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-463 Supplier Abnova Corporation Supplier No. MAB5793
Add to Cart
Add to Cart

Hamster monoclonal antibody raised against native Cd81.

Specifications

Antigen Cd81
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation
Classification Monoclonal
Clone 2F7
Conjugate PE
Description Hamster monoclonal antibody raised against native Cd81.
Dilution Flow Cytometry (0.2 ug/106 cells) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene Cd81
Gene Alias Tapa-1/Tapa1/Tspan28
Gene Symbols Cd81
Host Species Hamster
Immunogen Native purified Cd81 from mouse epithelial cell line PAM212.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12520
Test Specificity Mouse CD81 (Mr 25 KDa), a member of the transmembrane 4 integral membrane protein family.
Target Species Mouse
Content And Storage Store in the dark at 4°C. Do not freeze.
Avoid prolonged exposure to light.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG3 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.