Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ctla4, Hamster, Clone: 9H10, Abnova™

Catalog No. 89111289 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-289 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-289 Supplier Abnova Corporation Supplier No. MAB3241
Add to Cart
Add to Cart

Hamster monoclonal antibody raised against recombinant Ctla4.

Specifications

Antigen Ctla4
Applications Flow Cytometry, Immunoprecipitation
Classification Monoclonal
Clone 9H10
Conjugate Unconjugated
Description Hamster monoclonal antibody raised against recombinant Ctla4.
Dilution Flow Cytometry (3 ug/106 cells) The optimal working dilution should be determined by the end user.
Formulation In 100mM BBS, pH 8.2
Gene Ctla4
Gene Alias Cd152/Ctla-4/Ly-56
Gene Symbols Ctla4
Host Species Hamster
Immunogen Recombinant human IgG1 fusion protein corresponding to mouse Ctla4.
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12477
Test Specificity Mouse CD152/CTLA-4 costimulatory receptor, Mr 69 KDa.
Target Species Mouse
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.