Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FASLG, Hamster, Clone: 4A5, Abnova™

Catalog No. 89111972 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-972 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-972 Supplier Abnova Corporation Supplier No. MAB4703

Hamster monoclonal antibody raised against recombinant FASLG.

The protein encoded by this gene is the ligand for FAS. Both are transmembrane proteins. Interaction of FAS with this ligand is critical in triggering apoptosis of some types of cells such as lymphocytes. Defects in this gene may be related to some cases of systemic lupus erythematosus (SLE). [provided by RefSeq

Specifications

Antigen FASLG
Applications ELISA, Flow Cytometry
Classification Monoclonal
Clone 4A5
Conjugate Unconjugated
Description Fas ligand (TNF superfamily, member 6)
Formulation Lyophilized from PBS
Gene FASLG
Gene Alias APT1LG1, CD178, CD95L, FASL, TNFSF6
Gene Symbols FASLG
Host Species Hamster
Immunogen Recombinant protein corresponding to human FASLG.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 356
Target Species Human
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.