Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lair1, Hamster, Clone: 113, Abnova™

Catalog No. 89109212 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-212 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-212 Supplier Abnova Corporation Supplier No. MAB10996

Armenian hamster monoclonal antibody raised against Lair1.

Specifications

Antigen Lair1
Applications Flow Cytometry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 113
Conjugate Unconjugated
Description Armenian hamster monoclonal antibody raised against Lair1.
Dilution The optimal working dilution should be determined by the end user.
Formulation In aqueous buffer (0.09% sodium azide)
Gene Lair1
Gene Alias 5133400O11Rik/BB115266/D7Bwg0421e/Lair-1/mLair-1
Gene Symbols Lair1
Host Species Hamster
Immunogen Mouse Lair1.
Purification Method Affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 52855
Target Species Mouse
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.