Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ly49c/f/h/i, Hamster, Clone: 14B11, Abnova™

Catalog No. 89136151 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-151 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-151 Supplier Abnova Corporation Supplier No. MAB6211
Add to Cart
Add to Cart

Hamster monoclonal antibody raised against native Ly49c/f/h/i.

Specifications

Antigen Ly49c/f/h/i
Applications Flow Cytometry
Classification Monoclonal
Clone 14B11
Conjugate Unconjugated
Description Hamster monoclonal antibody raised against native Ly49c/f/h/i.
Dilution Flow Cytometry (1 ug/106 cells) The optimal working dilution should be determined by the end user.
Formulation In 100mM BBS, pH 8.2
Host Species Hamster
Immunogen Native purified from IL-2-activated killer cells (LAK) from C57BL/6 mice.
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity An epitope common to the Ly49C, Ly49F, Ly49H and Ly49I allelic/splice variants.
Target Species Mouse
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.