Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD154 Hamster anti-Mouse, Brilliant Violet 786, Clone: MR1, BD Optibuild™

Catalog No. BDB740874 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB740874 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB740874 Supplier BD Biosciences Supplier No. 740874
Add to Cart
Add to Cart

Hamster Monoclonal Antibody

The MR1 monoclonal antibody specifically binds to CD154 (CD40 Ligand, gp39), an accessory molecule expressed on activated T helper (CD4+) lymphocytes. CD154 has also been detected on other types of leukocytes, including CD8+ T cells, medullary thymocytes, activated CD4+ NK-T cells, and human NK cells. CD154 plays an important role in costimulatory interactions between T and B lymphocytes and between antigen-presenting cells and lymphocytes, regulating the immune response at multiple levels. MR1 mAb inhibits in vitro activation of B lymphocytes by T helper cells by blocking interaction of gp39 with CD40. In vitro interactions of T cells and antigen-presenting cells can also be blocked by the MR1 antibody. In vivo treatment with MR1 antibody blocks the development of experimental autoimmune disease, inhibits formation of germinal centers and generation of memory B cells, reduces T-lymphocyte responses to allogeneic cells and allografts, prevents intrathymic deletion of self-reactive T lymphocytes, and disrupts antigen-specific T-cell responses.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD154
Applications Flow Cytometry
Classification Monoclonal
Clone MR1
Concentration 0.2mg/mL
Conjugate Brilliant Violet 786
Description CD40 Ligand; CD40L; gp39; Ly-62; Tnfsf5; T-BAM; HIGM1; IMD3
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Hamster
Immunogen Activated mouse Th1 clone D1.6
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG3 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.