Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD272 Hamster anti-Mouse, Brilliant Violet 711, Clone: HMBT-6B2, BD Optibuild™

Catalog No. BDB740834 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB740834 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB740834 Supplier BD Biosciences Supplier No. 740834
Add to Cart
Add to Cart

Hamster Monoclonal Antibody

The HMBT-6B2 monoclonal antibody specifically binds to CD272 which is also known as B- and T-lymphocyte attenuator (BTLA). CD272 is a type 1 transmembrane glycoprotein and a member of the Ig superfamily. CD272 is variably expressed on developing and mature T and B lymphocytes, NKT cells, NK cells, macrophages and dendritic cells. CD272 expression is upregulated by activated T cells including Th1, Th2, and T follicular helper cells and by anergic T cells. CD272 is structurally similar to CD152/CTLA-4 and CD279/PD-1 and functions as a coinhibitory receptor for B and T cell responses. Herpesvirus entry mediator (HVEM), also known as CD270 and LIGHT-R, has been identified as a ligand for CD272. The crosslinking of CD272 by HVEM inhibits T-cell proliferation and cytokine production.

The antibody was conjugated to BD Horizon™ BV711 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 711-nm. BD Horizon BV711 can be excited by the violet laser and detected in a filter used to detect Cy™5.5 / Alexa Fluor™ 700-like dyes (eg, 712/20-nm filter). Due to the excitation and emission characteristics of the acceptor dye, there may be moderate spillover into the Alexa Fluor™ 700 and PerCP-Cy5.5 detectors. However, the spillover can be corrected through compensation as with any other dye combination.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD272
Applications Flow Cytometry
Classification Monoclonal
Clone HMBT-6B2
Concentration 0.2mg/mL
Conjugate Brilliant Violet 711
Description Btla; B and T lymphocyte attenuator; B- and T-lymphocyte-associated protein
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Hamster
Immunogen Mouse CD272
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.