Learn More
CD3e Hamster anti-Mouse, Brilliant Violet 786, Clone: 500A2, BD Optibuild™
Hamster Monoclonal Antibody
Supplier: BD Biosciences 740854
Description
The 500A2 monoclonal antibody specifically binds to the 25-kDa ε chain of the T-cell receptor-associated CD3 complex expressed on mouse thymocytes, mature T lymphocytes, and NKT cells. Plate-bound and soluble forms of the 500A2 antibody can activate T cells in vitro . Activation of a mouse T-cell clone by the 500A2 antibody can be blocked by Fab fragments of the GK1.5 anti-CD4 antibody. This suggests that the 500A2 antibody may bind an epitope on CD3e close to a site at which CD4 associates with the T-cell receptor. The 500A2 antibody reportedly does not to crossreact with rat leukocytes.

Specifications
CD3e | |
Monoclonal | |
0.2mg/mL | |
CD3; CD3 epsilon; Cd3e; CD3ε; T3e | |
Hamster | |
Affinity Purified | |
RUO | |
Mouse | |
IgG2 κ |
Flow Cytometry | |
500A2 | |
Brilliant Violet 786 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse T-cell receptor | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only